missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C3 Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8695-ec-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_000055.2 | |
Complement C3 Protein | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
YSIITPNILR LESEETMVLE AHDAQGDVPV TVTVHDFPGK KLVLSSEKTV LTPATNHMGN VTFTIPANRE FKSEKGRNKF VTVQATFGTQ VVEKVVLVSL QSGYLFIQTD KTIYTPGSTV LYRIFTVNHK LLPVGRTVMV NIENPEGIPV KQDSLSSQNQ LGVLPLSWDI PELVNMGQWK IRAYYENSPQ QVFSTEFEVK | |
Greater than 90% as determined by SDS-PAGE. |
718 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |