missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8456-ec-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_006821.1 | |
Cytochrome b-c1 complex subunit 10 Protein | |
Research Use Only | |
UQCR/UQCR11 | |
Recombinant Protein | |
E. coli | |
MVTRFLGPRY RELVKNWVPT AYTWGAVGAV GLVWATDWRL ILDWVPYING KFKKDN | |
Greater than 90% as determined by SDS-PAGE. |
10975 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |