missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D4GDI Protein
A cDNA sequence encoding the D4GDI was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11594-2ug
This item is not returnable.
View return policy
Specifications
D4GDI Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
D4-GDI protein at 0.1 mg/ml in 50mM Tris-HCl, pH 7.5 and 10mM L-Glutathione (reduced). |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 90% as determined by SDS-PAGE. |