missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Diacylglycerol kinase gamma Protein
A DNA sequence encoding the Homo sapiens (Human) Diacylglycerol kinase gamma, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5931-ec-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001074213.1 | |
Diacylglycerol kinase gamma Protein | |
Research Use Only | |
DGKG | |
Recombinant Protein | |
E. coli | |
MGEERWVSLT PEEFDQLQKY SEYSSKKIKD ALTEFNEGGS LKQYDPHEPI SYDVFKLFMR AYLEVDLPQP LSTHLFLAFS QKPRHETSDH PTEGASNSEA NSADTNIQNA DNATKADEAC APDTESNMAE KQAPAEDQVA ATPLEPPVPR SSSSESPVVY LKDVVCYLSL LETGRPQDKL EFMFRLYDSD ENGLLDQAEM DCIVNQMLHI AQYLEWDPTE LRPILKEMLQ GMDYDRDGFV SLQEWVHGGM TTIPL | |
Greater than 90% as determined by SDS-PAGE. |
1608 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |