missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENO1 / Enolase 1 / alpha-enolase Protein
A DNA sequence encoding the Homo sapiens (Human) ENO1 / Enolase 1 / alpha-enolase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8407-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001188412.1 | |
ENO1 / Enolase 1 / alpha-enolase Protein | |
Research Use Only | |
ENO1 | |
Recombinant Protein | |
E. coli | |
SILKIHAREI FDSRGNPTVE VDLFTSKGLF RAAVPSGAST GIYEALELRD NDKTRYMGKG VSKAVEHINK TIAPALVSKK LNVTEQEKID KLMIEMDGTE NKSKFGANAI LGVSLAVCKA GAVEKGVPLY RHIADLAGNS EVILPVPAFN VINGGSHAGN KLAMQEFMIL PVGAANFREA MRIGAEVYHN LKNVIKEKYG KDATNVGDEG GFAPNILENK EGLELLKTAI GKAGYTDKVV IGMDVAASEF FRSGKYDLDF KSPDDPSRYI SPDQLADLYK SFIKDYPVVS IEDPFDQDDW GAWQKFTASA GIQVVGDDLT VTNPKRIAKA VNEKSCNCLL LKVNQIGSVT ESLQACKLAQ ANGWGVMVSH RSGETEDTFI ADLVVGLCTG QIKTGAPCRS ERLAKYNQLL RIEEELGSKA KFAGRNF | |
Greater than 90% as determined by SDS-PAGE. |
2023 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |