missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epiregulin / EREG Protein
A DNA sequence encoding the Human Epiregulin / EREG, was expressed in the hosts and tags indicated.
105.00€ - 1970.00€
Especificaciones
Número de acceso | NP_001423.1 |
---|---|
ID de gen (Entrez) | 2069 |
Nombre | Epiregulin / EREG Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15933728
|
enQuireBio™
QP10397-ec-5ug |
5 μg | |||||||
15923728
|
enQuireBio™
QP10397-ec-25ug |
25 μg | |||||||
15903728
|
enQuireBio™
QP10397-ec-100ug |
100 μg | |||||||
15913728
|
enQuireBio™
QP10397-ec-250ug |
250 μg | |||||||
Especificaciones
NP_001423.1 | |
Epiregulin / EREG Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VAQVSITKCS SDMNGYCLHG QCIYLVDMSQ NYCRCEVGYT GVRCEHFFL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
2069 | |
Research Use Only | |
EREG | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |