Learn More
R&D Systems™ Recombinant Human FGF-8f Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 5027-FF-025/CF
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human FGF-8f Protein is derived from E. coli. The Recombinant Human FGF-8f Protein has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. | |
25.5 kDa | |
25μg | |
12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. | |
Recombinant |
2253 | |
FGF-8 | |
E. coli-derived human FGF-8 protein Gln23-Arg244,with an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |