Learn More
R&D Systems™ Recombinant Human FGF-8f Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca:  R&D Systems™ 5027-FF-025/CF
Código de producto. 18176910
  • 409.00€ / 25 microgramos
Explore más ofertas especiales
Descripción
Descripción
The Recombinant Human FGF-8f Protein is derived from E. coli. The Recombinant Human FGF-8f Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
25.5 kDa
25μg
12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution.
Recombinant
2253
FGF-8
E. coli-derived human FGF-8 protein Gln23-Arg244,with an N-terminal Met
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.