missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2 Protein, Animal-Free

Código de producto. 18786265
Click to view available options
Cantidad:
1 mg
25 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18786265

Marca: R&D Systems™ BTFGFBAFL01M

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Intended for preclinical researchers who may transition to GMP FGF basic/FGF2 for their clinical work

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.100-1.00 ng/mL.The specific activity of Recombinant Human FGF basic/FGF2 is >8.0 x 105 IU/mg, which is calibrated against the human FGF basic/FGF2 WHO International Standard (NIBSC code: 90/712).
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso NP_001997.5
Para utilizar con (aplicación) Bioactivity
Formulación Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
ID de gen (Entrez) 2247
Peso molecular M.W. (Observed): 18 kDa, under reducing conditions., M.W. (Theroretical): 16 kDa
Cantidad 1 mg
Fuente E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 Produced using non-animal reagents in an animal-free laboratory.
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Concentración de endotoxinas <0.01 EU per 1 μg of the protein by the LAL method.
Alias de gen Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Conjugado Unconjugated
Reconstitución Reconstitute 25 μg size at 250 μg/mL and others at 500 μg/mL in sterile water.
Grado de pureza o calidad >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Expresión FGF basic/FGF2/bFGF
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.