missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FUBP1 Protein
A cDNA sequence encoding the FUBP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11908-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
FUBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDVPIP RFAVGIVIGR NGEMIKKIQN DAGVRIQFKP DDGTTPERIA QITGPPDRCQ HAAEIITDLL RSVQAGNPGG PGPGGRGRGR GQGNWNMGPP GGLQEFNFIV PTGKTGLIIG KGGETIKSIS QQSGARIELQ RNPPPNADPN MKLFTIRGTP QQIDYARQLI EEKIG | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FUBP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.15M NaCl and 10% glycerol. |