missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein
A DNA sequence encoding the Human GM-CSF / CSF2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10346-ec-250ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_000749.2 | |
GM-CSF / CSF2 Protein | |
Research Use Only | |
CSF2 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
1437 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APARSPSPST QPWEHVNAIQ EARRLLNLSR DTAAEMNETV EVISEMFDLQ EPTCLQTRLE LYKQGLRGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFESFKENL KDFLLVIPFD CWEPVQE | |
Greater than 98% as determined by SDS-PAGE and HPLC. |