missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAI2 Protein
A cDNA sequence encoding the GNAI2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12010-5ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
GNAI2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGCTVSAEDK AAAERSKMID KNLREDGEKA AREVKLLLLG AGESGKSTIV KQMKIIHEDG YSEEECRQYR AVVYSNTIQS IMAIVKAMGN LQIDFADPSR ADDARQLFAL SCTAEEQGVL PDDLSGVIRR LWADHGVQAC FGRSREYQLN DSAAYYLNDL ERIAQSDYIP TQQDVLRTRV KTTGIVETHF TFKDLHFKMF DVGGQRSERK KWIHCFEGVT AIIFCVALSA YDLVLAEDEE MNRMHESMKL FDSICNNKWF TDTSIILFLN KKDLFEEKIT HSPLTICFPE YTGANKYDEA ASYIQSKFED LNKRKDTKEI YTHFTCATDT KNVQFVFDAV TDVIIKNNLK DCGLF | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GNAI2 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |