Learn More
enQuireBio™ Recombinant Human GPD2 Protein
A cDNA sequence encoding the GPD2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12037-2ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
2820 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAFQKAVKGT ILVGGGALAT VLGLSQFAHY RRKQMNLAYV KAADCISEPV NREPPSREAQ LLTLQNTSEF DILVIGGGAT GSGCALDAVT RGLKTALVER DDFSSGTSSR STKLIHGGVR YLQKAIMKLD IEQYRMVKEA LHERANLLEI APHLSAPLPI MLPVYKWWQL PYYWVGIKLY DLVAGSNCLK SSYVLSKSRA LEHFPMLQKD KLVGAIVYYD GQHNDARMNL AIALTAARYG AATANYMEVV SLLKKTDPQT GKVRVSGARC KDVLTGQEFD VRAKCVINAT GPFTDSVRKM DDKDAAAICQ PSAGVHIVMP GYYSPESMGL LDPATSDGRV IFFLPWQKMT IAGTTDTPTD VTHHPIPSEE DINFILNEVR NYLSCDVEVR RGDVLAAWSG IRPLVTDPKS ADTQSISRNH VVDISESGLI TIAGGKWTTY RSMAEDTINA AVKTHNLKAG PSRTVGLFLQ GGKDWSPTLY IRLVQDYGLE SEVAQHLAAT YGDKAFEVAK MASVTGKRWP IVGVRLVSEF PYIEAEVKYG IKEYACTAVD MISRRTRLAF LNVQAAEEAL PRIVELMGRE LNWDDYKKQE QLETARKFLY YEMGYKSRSE QLTDRSEISL LPSDIDRYKK RFHKFDADQK GFITIVDVQR VLESINVQMD ENTLHEILNE VDLNKNGQVE LNEFLQLMSA IQKGRVSGSR LAILMKTAEE NLDRRVPIPV DRSCGGL | |
Greater than 95.0% as determined by SDS-PAGE. |
GPD2 Protein | |
Research Use Only | |
GPD2 | |
Human | |
His | |
GPD2 protein solution is supplied in 20mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol. |