missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HDGF Protein
Highly purified. Generating reliable and reproducible results.
Marca: Novus Biologicals™ NBP1-30260
Detalles adicionales : Peso : 0.00970kg
Descripción
An un-tagged recombinant protein corresponding to the amino acids 1-100 of Human HDGF The Recombinant Human HDGF Protein is derived from E. coli. The Recombinant Human HDGF Protein has been validated for the following applications: SDS-Page.Especificaciones
NP_004485 | |
20mM Tris-HCl buffer (pH7.5), 1mM DTT, 10% glycerol, 1mM EDTA | |
11.5kDa | |
Protein | |
−80°C. Avoid freeze-thaw cycles. | |
>95%, by SDS-PAGE |
ELISA, SDS-PAGE | |
3068 | |
HDGF Protein | |
MSRSNRQKEY KCGDLVFAKM KGYPHWPARI DEMPEAAVKS TANKYQVFFF GTHETAFLGP KDLFPYEESK EKFGKPNKRK GFSEGLWEIE NNPTVKASGY | |
Human |