missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNA4 / Interferon alpha-4 Protein
A DNA sequence encoding the Homo sapiens (Human) IFNA4 / Interferon alpha-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6202-ec-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_066546.1 | |
IFNA4 / Interferon alpha-4 Protein | |
Research Use Only | |
IFNA4 | |
Recombinant Protein | |
E. coli | |
CDLPQTHSLG NRRALILLAQ MGRISHFSCL KDRHDFGFPE EEFDGHQFQK AQAISVLHEM IQQTFNLFST EDSSAAWEQS LLEKFSTELY QQLNDLEACV IQEVGVEETP LMNEDSILAV RKYFQRITLY LTEKKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD | |
Greater than 90% as determined by SDS-PAGE. |
3441 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |