Learn More
enQuireBio™ Recombinant Human IL13 / ALRH Protein
A DNA sequence encoding the Human IL13 / ALRH, was expressed in the hosts and tags indicated.
105.00€ - 3420.00€
Especificaciones
Número de acceso | NP_002179.2 |
---|---|
ID de gen (Entrez) | 3596 |
Nombre | IL13 / ALRH Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15983488
|
enQuireBio™
QP10320-ec-2ug |
2 μg | N/A | ||||||
15963488
|
enQuireBio™
QP10320-ec-10ug |
10 μg | N/A | ||||||
15953488
|
enQuireBio™
QP10320-ec-100ug |
100 μg | N/A | ||||||
15973488
|
enQuireBio™
QP10320-ec-250ug |
250 μg | N/A | ||||||
Especificaciones
NP_002179.2 | |
IL13 / ALRH Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPVPPSTALR ELIEELVNIT QNQKAPLCNG SMVWSINLTA GMYCAALESL INVSGCSAIE KTQRMLSGFC PHKVSAGQFS SLHVRDTKIE VAQFVKDLLL HLKKLFREGQ FN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
3596 | |
Research Use Only | |
IL13 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.2, with 5 % trehalose, lyophilized |