missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IMMP2L Protein
A cDNA sequence encoding the IMMP2L was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12430-20ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
IMMP2L Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRVEGASM QPSLNPGGSQ SSDVVLLNHW KVRNFEVHRG DIVSLVSPKN PEQKIIKRVI ALEGDIVRTI GHKNRYVKVP RGHIWVEGDH HGHSFDSNSF GPVSLGLLHA HATHILWPPE RWQKLESVLP PERLPVQREE E | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IMMP2L protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol and 1mM DTT. |