missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Insulysin/IDE Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marca: R&D Systems™ 2496-ZN-010
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPPGFSAFK(Dnp)-OH (Catalog # ES005 ). The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.Especificaciones
P14735 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, Brij-35 and Glycerol. | |
M.W. (Observed): 105 kDa, reducing conditions; M.W. (theoretical): 114 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
IDE | |
Recombinant | |
Insulysin/IDE |
Enzyme Activity | |
3416 | |
Insulysin/IDE | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Insulysin/IDE protein Met42-Leu1019, with an N-terminal Met and 7-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido