missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIR2DL5A Protein

A cDNA sequence encoding the KIR2DL5A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12490-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15900589

  • 212.00€ / 10 microgramos
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

KIR2DL5A Protein
Research Use Only
Recombinant Protein
Baculovirus
ADPHEGGQDK PLLSAWPSAV VPRGGHVTLL CRSRLGFTIF SLYKEDGVPV PELYNKIFWK SILMGPVTPA HAGTYRCRGS HPRSPIEWSA PSNPLVIVVT GLFGKPSLSA QPGPTVRTGE NVTLSCSSRS SFDMYHLSRE GRAHEPRLPA VPSVNGTFQA DFPLGPATHG GTYTCFGSLH DSPYEWSDPS DPLLVSVTGN SSSSSSSPTE PSSKTGIRRH VEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGKHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
KIR2DL5A protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales