missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIR2DL5A Protein
A cDNA sequence encoding the KIR2DL5A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12490-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
KIR2DL5A Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPHEGGQDK PLLSAWPSAV VPRGGHVTLL CRSRLGFTIF SLYKEDGVPV PELYNKIFWK SILMGPVTPA HAGTYRCRGS HPRSPIEWSA PSNPLVIVVT GLFGKPSLSA QPGPTVRTGE NVTLSCSSRS SFDMYHLSRE GRAHEPRLPA VPSVNGTFQA DFPLGPATHG GTYTCFGSLH DSPYEWSDPS DPLLVSVTGN SSSSSSSPTE PSSKTGIRRH VEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGKHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KIR2DL5A protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |