Learn More
R&D Systems™ Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 629-LR-100
404.31 EUR hasta el 2024-12-16
Use el código promocional "21630" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera is derived from NS0. The Recombinant Human Lymphotoxin beta R (LTbR) Fc Chimera has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS. | |
49.6 kDa (monomer) | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
4055 | |
Lymphotoxin beta R/TNFRSF3 Fc Chimera | |
Mouse myeloma cell line,NS0-derived human Lymphotoxin beta R/TNFRSF3 protein, (N-terminus) Human LT beta R (Ser28-Met227) & (Gln31-Met227) Accession &Num; NP_002333 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
Corrección del contenido de un producto