missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MANF / ARMET Protein
A DNA sequence encoding the Human MANF / ARMET, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10418-ec-5ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_006001.4 | |
MANF / ARMET Protein | |
Research Use Only | |
MANF | |
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 20 ug/ml, corresponding to a specific activity of > 50 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
7873 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIENELIKFC REARGKENRL CYYIGATDDA ATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKIL DDWGETCKGC AEKSDYIRKI NELMPKYAPK AASARTDL | |
Greater than 95% as determined by SDS-PAGE and HPLC. |