missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nicotinate phosphoribosyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Nicotinate phosphoribosyltransferase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6403-ye-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001273758.1 | |
Nicotinate phosphoribosyltransferase Protein | |
Research Use Only | |
NAPRT1 | |
Recombinant Protein | |
Yeast | |
MLAPAAGEGP GVDLAAKAQV WLEQVCAHLG LGVQEPHPGE RAAFVAYALA FPRAFQGLLD TYSVWRSGLP NFLAVALALG ELGYRAVGVR LDSGDLLQQA QEIRKVFRAA AAQFQVPWLE SVLIVVSNNI DEEALARLAQ EGSEVNVIGI GTSVVTCPQQ PSLGGVYKLV AVGGQPRMKL TEDPEKQTLP GSKAAFRLLG SDGSPLMDML QLAEEPVPQA GQELRVWPPG AQEPCTVRPA QVEPLLRLCL QQGQLCEPLP SLAESRALAQ LSLSRLSPEH RRLRSPAQYQ VVLSERLQAL VNSLCAGQSP | |
Greater than 90% as determined by SDS-PAGE. |
93100 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |