missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear RNA export factor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear RNA export factor 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6443-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_071335.1 | |
Nuclear RNA export factor 3 Protein | |
Research Use Only | |
NXF3 | |
Recombinant Protein | |
E. coli | |
MSLPSGHTTG HTDQVVQRRA RCWDIYQRRF SSRSEPVNPG MHSSSHQQQD GDAAMHGAHM DSPVRYTPYT ISPYNRKGSF RKQDQTHVNM EREQKPPERR MEGNMPDGTL GSWFKITVPF GIKYNEKWLL NLIQNECSVP FVPVEFHYEN MHASFFVENA SIAYALKNVS GKIWDEDNEK ISIFVNPAGI PHFVHRELKS EKVEQIKLAM NQQCDVSQEA LDIQRLPFYP DMVNRDTKMA SNPRKCMAAS LDVHEENIPT VMSAGEMDKW KGIEPGEKCA DRSPVCTTFS DTSSNINSIL ELFPKLLCLD GQQSPRATLC GTEAHKRLPT CKGSFFGSEM LKNLVLQFLQ QYYLIYDSGD RQGLLSAYHD EACFSLSIPF NPEDSAPSSF CKFFKDSRNI KILKDPYLRG ELLKHTKLDI VDSLSALPKT QHDLSSFLVD MWYQTEWMLC FSVNGVFKEV EGQSQGSVLA FTRTFIATPG SSSSLCIVND KLFVRDTSHQ GTQSALFTLV PTAFSSSVPA FSQEQQKMLP S | |
Greater than 90% as determined by SDS-PAGE. |
56000 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |