missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Pax5/BSAP GST (N-Term) Protein
Recombinant Protein
Marca: Novus Biologicals™ H00005079-Q01-25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Especificaciones
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
5079 | |
25 μg | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
Cancer, Transcription Factors and Regulators | |
Pax5/BSAP |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
37.84 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
PAX5 | |
>80% by SDS-PAGE and Coomassie blue staining |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido