missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase-like 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase-like 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7976-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_624311.1 | |
Peptidyl-prolyl cis-trans isomerase-like 4 Protein | |
Research Use Only | |
PPIL4 | |
Recombinant Protein | |
E. coli | |
MAVLLETTLG DVVIDLYTEE RPRACLNFLK LCKIKYYNYC LIHNVQRDFI IQTGDPTGTG RGGESIFGQL YGDQASFFEA EKVPRIKHKK KGTVSMVNNG SDQHGSQFLI TTGENLDYLD GVHTVFGEVT EGMDIIKKIN ETFVDKDFVP YQDIRINHTV ILDDPFDDPP DLLIPDRSPE PTREQLDSGR IGADEEIDDF KGRSAEEVEE IKAEKEAKTQ AILLEMVGDL PDADIKPPEN VLFVCKLNPV TTDEDLEIIF SRFGPIRSCE VIRDWKTGES LCYAFIEFEK EEDCEKAFFK MDNVLIDDRR IHVDFSQSVA KVKWKGKGGK YTKSDFKEYE KEQDKPPNLV LKDKVKPKQD TKYDLILDEQ AEDSKSSHSH TSKKHKKKTH HCSEEKEDED YMPIKNTNQD IYREMGFGHY EEEESCWEKQ KSEKRDRTQN RSRSRSRERD GHYSNSHKSK YQTDLYERER SKKRDRSRSP KKSKDKEKSK YR | |
Greater than 90% as determined by SDS-PAGE. |
85313 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |