missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRDX3 Protein
A cDNA sequence encoding the PRDX3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10839-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
PRDX3 Protein | |
Research Use Only | |
The specific activity was found to be approximately 82-83 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX3 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute. | |
Human | |
Untagged | |
The PRDX3 solution contains 20mM Tris-HCl pH 8.0, and 10% glycerol. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MPAVTQHAPY FKGTAVVNGE FKDLSLDDFK GKYLVLFFYP LDFTFVCPTE IVAFSDKANE FHDVNCEVVA VSVDSHFSHL AWINTPRKNG GLGHMNIALL SDLTKQISRD YGVLLEGSGL ALRGLFIIDP NGVIKHLSVN DLPVGRSVEE TLRLVKAFQY VETHGEVCPA NWTPDSPTIK PSPAASKEYF QKVNQ | |
Greater than 95.0% as determined by SDS-PAGE. |