missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable E3 SUMO-protein ligase RNF212 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable E3 SUMO-protein ligase RNF212, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6612-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001124506.1 | |
Probable E3 SUMO-protein ligase RNF212 Protein | |
Research Use Only | |
RNF212 | |
Recombinant Protein | |
E. coli | |
MANWVFCNRC FQPPHRTSCF SLTNCGHVYC DACLGKGKKN ECLICKAPCR TVLLSKHTDA DIQAFFMSID SLCKKYSRET SQILEFQEKH RKRLLAFYRE KISRLEESLR KSVLQIEQLQ SMRSSQQTAF STIKSSVSTK PHGCLLPPHS SAPDRLESME VDLSPSPIRK SEIAAGPARI SMISPPQDGR MGPHLTASFC FIPWLTLSKP PVPGECVISR GSPCFCIDVC PHWLLLLAFS SGRHGELTNS KTLPIYAEVQ RAVLFPFQQA EGTLDTFRTP AVSVVFPLCQ FERKKSF | |
Greater than 80% as determined by SDS-PAGE. |
285498 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |