missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein E6 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7409-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_041325.1 | |
Protein E6 Protein | |
Research Use Only | |
E6 | |
Recombinant Protein | |
E. coli | |
MHQKRTAMFQ DPQERPRKLP QLCTELQTTI HDIILECVYC KQQLLRREVY DFAFRDLCIV YRDGNPYAVC DKCLKFYSKI SEYRHYCYSL YGTTLEQQYN KPLCDLLIRC INCQKPLCPE EKQRHLDKKQ RFHNIRGRWT GRCMSCCRSS RTRRETQL | |
Greater than 90% as determined by SDS-PAGE. |
1489078 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |