missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RALGPS1 Protein
A DNA sequence encoding the Homo sapiens (Human) RALGPS1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7659-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001177657.1 | |
RALGPS1 Protein | |
Research Use Only | |
RALGPS1 | |
Recombinant Protein | |
E. coli | |
MYKRNGLMAS VLVTSATPQG SSSSDSLEGQ SCDYASKSYD AVVFDVLKVT PEEFASQITL MDIPVFKAIQ PEELASCGWS KKEKHSLAPN VVAFTRRFNQ VSFWVVREIL TAQTLKIRAE ILSHFVKIAK KLLELNNLHS LMSVVSALQS APIFRLTKTW ALLNRKDKTT FEKLDYLMSK EDNYKRTREY IRSLKMVPSI PYLGIYLLDL IYIDSAYPAS GSIMENEQRS NQMNNILRII ADLQVSCSYD HLTTLPHVQK YLKSVRYIEE LQKFVEDDNY KLSLRIEPGS SSPRLVSSKE DLAAM | |
Greater than 90% as determined by SDS-PAGE. |
9649 | |
500 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |