missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RELA / Transcription factor p65 / NFkB p65 Protein
A DNA sequence encoding the Homo sapiens (Human) RELA / Transcription factor p65 / NFkB p65, was expressed in the hosts and tags indicated.
160.00€ - 1855.00€
Especificaciones
Número de acceso | NP_001138610.1 |
---|---|
ID de gen (Entrez) | 5970 |
Nombre | RELA / Transcription factor p65 / NFkB p65 Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Not determined. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15912849
|
enQuireBio™
QP1321-ec-10ug |
10 μg | |||||||
15952849
|
enQuireBio™
QP1321-ec-50ug |
50 μg | |||||||
15902849
|
enQuireBio™
QP1321-ec-100ug |
100 μg | |||||||
15932849
|
enQuireBio™
QP1321-ec-200ug |
200 μg | |||||||
15942849
|
enQuireBio™
QP1321-ec-500ug |
500 μg | |||||||
15922849
|
enQuireBio™
QP1321-ec-1mg |
1 mg | |||||||
Especificaciones
NP_001138610.1 | |
RELA / Transcription factor p65 / NFkB p65 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5970 | |
Research Use Only | |
RELA | |
Recombinant Protein | |
E. coli | |
MDELFPLIFP AEPAQASGPY VEIIEQPKQR GMRFRYKCEG RSAGSIPGER STDTTKTHPT IKINGYTGPG TVRISLVTKD PPHRPHPHEL VGKDCRDGFY EAELCPDRCI HSFQNLGIQC VKKRDLEQAI SQRIQTNNNP FQVPIEEQRG DYDLNAVRLC FQVTVRDPSG RPLRLPPVLS HPIFDNRAPN TAELKICRVN RNSGSCLGGD | |
Greater than 90% as determined by SDS-PAGE. |