missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribonuclease P protein subunit p20 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonuclease P protein subunit p20, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6515-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005828.2 | |
Ribonuclease P protein subunit p20 Protein | |
Research Use Only | |
POP7 | |
Recombinant Protein | |
E. coli | |
MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINR AINIALQLQA GSFGSLQVAA NTSTVELVDE LEPETDTREP LTRIRNNSAI HIRVFRVTPK | |
Greater than 80% as determined by SDS-PAGE. |
10248 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |