missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribonuclease UK114 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonuclease UK114, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6182-ec-100ug
Additional Details : Peso : 0.01000kg
Specifications
NP_005827.1 | |
Ribonuclease UK114 Protein | |
Research Use Only | |
HRSP12 | |
Recombinant Protein | |
E. coli | |
SSLIRRVIST AKAPGAIGPY SQAVLVDRTI YISGQIGMDP SSGQLVSGGV AEEAKQALKN MGEILKAAGC DFTNVVKTTV LLADINDFNT VNEIYKQYFK SNFPARAAYQ VAALPKGSRI EIEAVAIQGP LTTASL | |
Greater than 90% as determined by SDS-PAGE. |
10247 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |