missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RRM1 Protein
A DNA sequence encoding the Homo sapiens (Human) RRM1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6629-ye-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001024.1 | |
RRM1 Protein | |
Research Use Only | |
RRM1 | |
Recombinant Protein | |
Yeast | |
MHVIKRDGRQ ERVMFDKITS RIQKLCYGLN MDFVDPAQIT MKVIQGLYSG VTTVELDTLA AETAATLTTK HPDYAILAAR IAVSNLHKET KKVFSDVMED LYNYINPHNG KHSPMVAKST LDIVLANKDR LNSAIIYDRD FSYNYFGFKT LERSYLLKIN GKVAERPQHM LMRVSVGIHK EDIDAAIETY NLLSERWFTH ASPTLFNAGT NRPQLSSCFL LSMKDDSIEG IYDTLKQCAL ISKSAGGIGV AVSCIRATGS YIAGTNGNSN GLVPMLRVYN NTARYVDQGG NKRPGAFAIY LEPWHLDIFE FLDLKKNTGK EEQRARDLFF ALWIPDLFMK RVETNQDWSL MCPNECPGLD EVWGEEFEKL YASYEKQGRV RKVVKAQQLW YAIIESQTET GTPYMLYKDS CNRKSNQQNL GTIKCSNLCT EIVEYTSKDE VAVCNLASLA LNMYVTSEHT YDFKKLAEVT KVVVRNLNKI IDINYYPVPE ACLSNKRHRP IGIGVQGLAD AFILMRYPFE SAEAQLLNKQ IFETIYYGAL EASCDLAKEQ GPYETYEGSP VSKGILQYDM WNVTPTDLWD WKVLKEKIAK YGIRNSLLIA PMPTASTAQI LGNNESIEPY TSNIYTRRVL SGEFQIVNPH LLKDLTERGL WHEEMKNQII ACNGSIQSIP EIPDDLKQLY KTVWEISQKT VLKMAAERGA FIDQSQSLNI HIAEPNYGKL TSMHFYGWKQ GLKTGMYYLR TRPAANPIQF TLNKEKLKDK EKVSKEEEEK ERNTAAMVCS LENRDECLMC GS | |
Greater than 90% as determined by SDS-PAGE. |
6240 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |