missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SARAF Protein
A DNA sequence encoding the Homo sapiens (Human) SARAF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7982-ec-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001271168.1 | |
SARAF Protein | |
Research Use Only | |
SARAF | |
Recombinant Protein | |
E. coli | |
SDGQYSPPPY SEYPPFSHRY QRFTNSAGPP PPGFKSEFTG PQNTGHGATS GFGSAFTGQQ GYENSGPGFW TGLGTGGILG YLFGSNRAAT PFSDSWYYPS YPPSYPGTWN RAYSPLHGGS GSYSVCSNSD TKTRTASGYG GTRRR | |
Greater than 90% as determined by SDS-PAGE. |
51669 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |