missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SF3B14 Protein
A cDNA sequence encoding the SF3B14 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13476-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
SF3B14 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAMQAAKRAN IRLPPEVNRI LYIRNLPYKI TAEEMYDIFG KYGPIRQIRV GNTPETRGTA YVVYEDIFDA KNACDHLSGF NVCNRYLVVL YYNANRAFQK MDTKKKEEQL KLLKEKYGIN TDPPK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SF3B14 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT, 1mM EDTA and 30% glycerol. |