Learn More
R&D Systems™ Recombinant Human sFRP-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca:  R&D Systems™ 1827-SF-025
 Ver más versiones de este producto
Código de producto. 18136280
  • 484.00€ / 25 microgramos
Explore más ofertas especiales
Descripción
Descripción
The Recombinant Human sFRP-4 Protein is derived from NS0. The Recombinant Human sFRP-4 Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
38.9 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
6424
sFRP-4
Mouse myeloma cell line,NS0-derived human sFRP-4 protein Ala22-Val346,with a C-terminal 10-His tag
Recombinant Human sFRP-4 is prone to proteolytic cleavage at C-terminus, therefore the poly-His tag at C-terminus may not be present in the preparation.
Recombinant
Sugerencias de productos