Learn More
R&D Systems™ Recombinant Human sFRP-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 1827-SF-025/CF
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human sFRP-4 Protein is derived from NS0. The Recombinant Human sFRP-4 Protein has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS. | |
38.9 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
6424 | |
sFRP-4 | |
Mouse myeloma cell line,NS0-derived human sFRP-4 protein Ala22-Val346,with a C-terminal 10-His tag | |
Recombinant human sFRP-4 is prone to proteolytic cleavage at C-terminus, therefore the poly-His tag at C-terminus may not be present in the preparation. | |
Recombinant |
Corrección del contenido de un producto