missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sorting nexin-20 Protein
A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-20, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6709-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001138444.1 | |
Sorting nexin-20 Protein | |
Research Use Only | |
SNX20 | |
Recombinant Protein | |
E. coli | |
MASPEHPGSP GCMGPITQCT ARTQQEAPAT GPDLPHPGPD GHLDTHSGLS SNSSMTTREL QQYWQNQKCR WKHVKLLFEI ASARIEERKV SKFVVYQIIV IQ | |
Greater than 90% as determined by SDS-PAGE. |
124460 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |