missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transglutaminase II Protein
A DNA sequence encoding the Homo sapiens (Human) Transglutaminase II, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6784-ec-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001310245.1 | |
Transglutaminase II Protein | |
Research Use Only | |
TGM2 | |
Recombinant Protein | |
E. coli | |
ELVLERCDLE LETNGRDHHT ADLCREKLVV RRGQPFWLTL HFEGRNYEAS VDSLTFSVVT GPAPSQEAGT KARFPLRDAV EEGDWTATVV DQQDCTLSLQ LTTPANAPIG LYRLSLEAST GYQGSSFVLG HFILLFNAWC PADAVYLDSE EERQEYVLTQ QGFIYQGSAK FIKNIPWNFG QFEDGILDIC LILLDVNPKF LKNAGRDCSR RSSPVYVGRV VSGMVNCNDD QGVLLGRWDN NYGDGVSPMS WIGSVDILRR WKNHGCQRVK YGQCWVFAAV ACTVLRCLGI PTRVVTNYNS AHDQNSNLLI EYFRNEFGEI QGDKSEMIWN FHCWVESWMT RPDLQPGYEG WQALDPTPQE KSEGTYCCGP VPVRAIKEGD LSTKYDAPFV FAEVNADVVD WIQQDDGSVH KSINRSLIVG LKISTKSVGR DEREDITHTY KYPEGSSEER EAFTRANHLN KLAEKEETGM AMRIRVGQSM NMGSDFDVFA HITNNTAEEY VCRLLLCART VSYNGILGPE CGTKYLLNLN LEPFSGKALC S | |
Greater than 90% as determined by SDS-PAGE. |
7052 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |