missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Homo sapiens (Human) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6770-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_005642.1 | |
Tryptophan 2,3-dioxygenase Protein | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
E. coli | |
MSGCPFLGNN FGYTFKKLPV EGSEEDKSQT GVNRASKGGL IYGNYLHLEK VLNAQELQSE TKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VVSRMHRVSV ILKLLVQQFS ILETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQN MRVPYNRRHY RDNFKGEENE LLLKSEQEKT LLELVEAWLE RTPGLEPHGF NFWGKLEKNI TRGLEEEFIR IQAKEESEEK EEQVAEFQKQ KEVLLSLFDE KRHEHLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDSL MTKWRYNHVC MVHRMLGSKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLIPRHWIPK MNPTIHKFLY TAEYCDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |
6999 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |