missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VCL / Vinculin Protein
A DNA sequence encoding the Homo sapiens (Human) VCL / Vinculin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8773-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_003364.1 | |
VCL / Vinculin Protein | |
Research Use Only | |
VCL | |
Recombinant Protein | |
E. coli | |
PVFHTRTIES ILEPVAQQIS HLVIMHEEGE VDGKAIPDLT APVAAVQAAV SNLVRVGKET VQTTEDQILK RDMPPAFIKV ENACTKLVQA AQMLQSDPYS VPARDYLIDG SRGILSGTSD LLLTFDEAEV RKIIRVCKGI LEYLTVAEVV ETMEDLVTYT KNLGPGMTKM AKMIDERQQE LTHQEHRVML VNSMNTVKEL LPVLISAMKI FVTTKNSKNQ GIEEALKNRN FTVE | |
Greater than 90% as determined by SDS-PAGE. |
7414 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |