missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human ZBTB7A/Pokemon GST (N-Term) Protein
Tienda Bio Techne ProductosDescripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulación | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| ID de gen (Entrez) | 51341 |
| Peso molecular | MolecularWeight-theroretical: 37.62 kDa |
| Cantidad | 10 μg |
| Requisitos de almacenamiento | Store at -80°C. Avoid freeze-thaw cycles. |
| Alias de gen | DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor |
| Símbolo de gen | ZBTB7A |
| Categoría de investigación | Cancer, Transcription Factors and Regulators |
| Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?