missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL13 / ALRH Protein
A DNA sequence encoding the Rhesus Macaque IL13 / ALRH, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10442-ec-250ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001028101.1 | |
IL13 / ALRH Protein | |
Research Use Only | |
IL13 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 5 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 3% trehalose, lyophilized |
574325 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPSPVPRSTA LKELIEELVN ITQNQKAPLC NGSMVWSINL TAGVYCAALE SLINVSGCSA IEKTQRMLNG FCPHKVSAGQ FSSLRVRDTK IEVAQFVKDL LVHLKKLFRE GRFN | |
Greater than 98% as determined by SDS-PAGE and HPLC. |