missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AFP / alpha-fetoProtein
A DNA sequence encoding the Mus musculus (Mouse) AFP / alpha-fetoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5619-ec-500ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_031449.3 | |
AFP / alpha-fetoProtein | |
Research Use Only | |
Afp | |
Recombinant Protein | |
E. coli | |
KALHENEFGI ASTLDSSQCV TEKNVLSIAT ITFTQFVPEA TEEEVNKMTS DVLAAMKKNS GDGCLESQLS VFLDEICHET ELSNKYGLSG CCSQSGVERH QCLLARKKTA PASVPPFQFP EPAESCKAHE ENRAVFMNRF IYEVSRRNPF MYAPAILSLA AQYDKVVLAC CKADNKEECF QTKRASIAKE LREGSMLNEH VCSVIRKFGS RNLQATTIIK LSQKLTEANF TEIQKLALDV AHIHEECCQG NSLECLQDGE KVMTYICSQQ NILSSKIAEC CKLPMIQLGF CIIHAENGVK PEGLSLNPSQ FLGDRNFAQF SSEEKIMFMA SFLHEYSRTH PNLPVSVILR IAKTYQEILE KCSQSGNLPG CQDNLEEELQ KHIEESQALS KQSCALYQTL GDYKLQNLFL IGYTRKAPQL TSAELIDLTG KMVSIASTCC QLSEEKWSGC GEGMADIFIG HLCIRNEASP VNSGISHCCN SSYSNRRLCI TSFLRDETYA PPPFSEDKFI FHKDLCQAQG KALQTMKQEL LINLVKQKPE LTEEQLAAVT ADFSGLLEKC CKAQDQEVCF TEEGPKLISK TRDALGV | |
Greater than 90% as determined by SDS-PAGE. |
11576 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |