missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD207 / Langerin Protein
A DNA sequence encoding the Mus musculus (Mouse) CD207 / Langerin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7887-ec-500ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_659192.2 | |
CD207 / Langerin Protein | |
Research Use Only | |
Cd207 | |
Recombinant Protein | |
E. coli | |
QAVFYPRLMG KILDVKSDAQ MLKGRVDNIS TLGSDLKTER GRVDDAEVQM QIVNTTLKRV RSQILSLETS MKIANDQLQI LTMSWGEVDS LSAKIPELKR DLDKASALNT KVQGLQNSLE NVNKLLKQQS DILEMVARGW KYFSGNFYYF SRTPKTWYSA EQFCISRKAH LTSVSSESEQ KFLYKAADGI PHWIGLTKAG SEGDWYWVDQ TSFNKEQSRR FWIPGEPNNA GNNEHCANIR VSALKCWNDG PCDNTFLFIC KRPYVQTTE | |
Greater than 80% as determined by SDS-PAGE. |
246278 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |