Learn More
R&D Systems™ Recombinant Mouse IGFBP-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 8066-GB-025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Mouse IGFBP-4 Protein is derived from NS0. The Recombinant Mouse IGFBP-4 Protein has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
26 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
16010 | |
IGFBP-4 | |
Mouse myeloma cell line,NS0-derived mouse IGFBP-4 protein Asp22-Glu254 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |