missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-5 R alpha Fc Chimera Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Marca:  R&D Systems™ 553-MR-025

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18117820

  • 356.36€ / 25 microgramos
Fecha estimada de envío: 06-08-2024
para ver el stock

Descripción

Descripción

The Recombinant Mouse IL-5 R alpha Fc Chimera Protein is derived from NS0. The Recombinant Mouse IL-5 R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.
Especificaciones

Especificaciones

Lyophilized from a 0.2μm filtered solution in PBS.
Observed Molecular Wt.: 75-85 kDa, 55kDa and 35kDa under reducing conditions
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
16192
IL-5R alpha/CD125
Mouse myeloma cell line,NS0-derived mouse IL-5 R alpha/CD125 protein, (N-terminus) Mouse IL-5 R alpha (Asp18-His339) Accession &Num; P21183 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales