missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL6 / Interleukin-6 Protein
Recombinant Protein
84.15€ - 3300.00€
Especificaciones
Número de acceso | NP_001300983.1 |
---|---|
ID de gen (Entrez) | 16193 |
Nombre | IL6 / Interleukin-6 Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15945489
|
enQuireBio™
QP1487-2 ug |
2 μg | |||||||
15925489
|
enQuireBio™
QP1487-10 ug |
10 μg | |||||||
15935489
|
enQuireBio™
QP1487-100 ug |
100 μg | |||||||
15955489
|
enQuireBio™
QP1487-500 ug |
500 μg | |||||||
15915489
|
enQuireBio™
QP1487-1 mg |
1 mg | |||||||
Especificaciones
NP_001300983.1 | |
IL6 / Interleukin-6 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the 7TD1 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
16193 | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
E. coli | |
MFPTSQVRRG DFTEDTTPNR PVYTTSQVGG LITHVLWEIV EMRKELCNGN SDCMNNDDAL AENNLKLPEI QRNDGCYQTG YNQEICLLKI SSGLLEYHSY LEYMKNNLKD NKKDKARVLQ RDTETLIHIF NQEVKDLHKI VLPTPISNAL LTDKLESQKE WLRTKTIQFI LKSLEEFLKV TLRSTRQT | |
Greater than 95% as determined by SDS-PAGE. |