missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Perforin-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Perforin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6526-ec-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_035203.3 | |
Perforin-1 Protein | |
Research Use Only | |
Prf1 | |
Recombinant Protein | |
E. coli | |
PCYTATRSEC KQKHKFVPGV WMAGEGMDVT TLRRSGSFPV NTQRFLRPDR TCTLCKNSLM RDATQRLPVA ITHWRPHSSH CQRNVAAAKV HSTEGVAREA AANINNDWRV GLDVNPRPEA NMRASVAGSH SKVANFAAEK TYQDQYNFNS DTVECRMYSF RLVQKPPLHL DFKKALRALP RNFNSSTEHA YHRLISSYGT HFITAVDLGG RISVLTALRT CQLTLNGLTA DEVGDCLNVE AQVSIGAQAS VSSEYKACEE KKKQHKMATS FHQTYRERHV EVLGGPLDST HDLLFGNQAT PEQFSTWTAS LPSNPGLVDY SLEPLHTLLE EQNPKREALR QAISHYIMSR ARWQNCSRPC RSGQHKSSHD SCQCECQDSK VTNQDCCPRQ RGLAHLVVSN FRAEHLWGDY TTATDAYLKV FFGGQEFRTG VVWNNNNPRW TDKMDFENVL LSTGGPLRVQ VWDADYGWDD DLLGSCDRSP HSGFHEVTCE LNHGRVKFSY HAKCLPHLTG GTCLEYAPQG LLGDPPGNRS GAVW | |
Greater than 80% as determined by SDS-PAGE. |
18646 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |