missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia OspA Protein

A cDNA sequence encoding the Borrelia OspA was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11189-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15996418

  • 212.00€ / 20 microgramos
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

Borrelia OspA Protein
Research Use Only
Recombinant Protein
E. coli
Borrelia OspA is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales